Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024936.hg.1 |
TC06002004.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024936.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024936.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202451.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202451.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202451.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202451.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202451.hg.1 |
TC0600014332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |