Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024074.hg.1 |
TC06001956.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024074.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024074.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024074.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012628.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
ENST00000316149.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
ENST00000478382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
ENST00000489477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
NM_014165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186984.hg.1 |
TC0600012628.hg.1 |
Clariom D TC |