Assay Details
| Gene Symbol: GNGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC029367.1 | - | 1 - 2 | 135 | 4604788 | 149 |
| BG435040.1 | - | 1 - 2 | 135 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC029367.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008340.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158272.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158272.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158272.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158272.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158272.hg.1 | TC0700008340.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | ENST00000455502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158278.hg.1 | TC0700008340.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000248572.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000428834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000429473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | ENST00000430875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | NM_021955.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158275.hg.1 | TC0700008340.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 100103 | - | - |

GNGT1
Genomic Map