Assay Details
| Gene Symbol: SLC6A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318369.1 | NP_001305298.1 | 1 - 2 | 270 | 100 | |
| NM_004211.4 | NP_004202.3 | 1 - 2 | 270 | 100 | ||
| GenBank mRNA | AF085412.1 | - | 1 - 2 | 278 | 100 | |
| BC096319.3 | - | 1 - 2 | 76 | 40007841 | 100 | |
| BC096320.3 | - | 1 - 2 | 76 | 40007842 | 100 | |
| BC096321.1 | - | 1 - 2 | 76 | 40007845 | 100 | |
| BC096322.3 | - | 1 - 2 | 76 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000258.hg
Additional Information:
This assay is in the 5'UTR region of the AF085412.1,BC096319.3,BC096320.3,BC096321.1,BC096322.3,NM_001318369.1,NM_004211.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003443.hg.1 | ENST00000525748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003443.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003443.hg.1 | TC11000258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003444.hg.1 | ENST00000525748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003444.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003444.hg.1 | TC11000258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003445.hg.1 | ENST00000525748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003445.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003445.hg.1 | TC11000258.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100149732.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149732.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149732.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149732.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149732.hg.1 | TC1100007055.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100149735.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149735.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149735.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149735.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149735.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149735.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149735.hg.1 | TC1100007055.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | ENST00000528440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007055.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149733.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149733.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149733.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149733.hg.1 | ENST00000298923.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149733.hg.1 | ENST00000525748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149733.hg.1 | NM_004211.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149733.hg.1 | TC1100007055.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 145340 | - | - |

SLC6A5
Genomic Map