Assay Details
| Gene Symbol: TYW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018264.3 | NP_060734.2 | 8 - 9 | 1279 | 77 | |
| XM_011516372.2 | XP_011514674.1 | 8 - 9 | 1267 | 77 | ||
| GenBank mRNA | AK001762.1 | - | 8 - 9 | 1246 | 77 | |
| BC015591.2 | - | 8 - 9 | 1245 | 4644035 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000414.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000359626.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000361660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000442959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000442959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000442959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000442959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000442959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000442959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000475392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000475392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000475392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000475392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000475392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000475392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000491969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000491969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000491969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000495971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | ENST00000615572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007859.hg.1 | NM_018264.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000610600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000612372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | ENST00000620995.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011463.hg.1 | NM_001145440.2 | Refseq |

TYW1
Genomic Map