Assay Details
| Gene Symbol: TK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172643.1 | NP_001166114.1 | 1 - 2 | 40 | 72 | |
| NM_001271935.1 | NP_001258864.1 | 1 - 2 | 40 | 72 | ||
| GenBank mRNA | U77088.1 | - | 1 - 2 | 40 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

TK2
Genomic Map