Assay Details
| Gene Symbol: CAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001752.3 | NP_001743.1 | 1 - 2 | 159 | 86 | |
| GenBank mRNA | AK291585.1 | - | 1 - 2 | 159 | 86 | |
| AK315350.1 | - | 1 - 2 | 143 | 86 | ||
| AY028632.1 | - | 1 - 2 | 70 | 86 | ||
| BC110398.1 | - | 1 - 2 | 148 | 5551309 | 86 | |
| BC112217.1 | - | 1 - 2 | 108 | 8327685 | 86 | |
| BC112219.1 | - | 1 - 2 | 108 | 8327687 | 86 | |
| X04076.1 | - | 1 - 2 | 140 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000327.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
osteoblast differentiation
ureteric bud development
response to hypoxia
purine nucleotide catabolic process
triglyceride metabolic process
aging
cholesterol metabolic process
aerobic respiration
response to light intensity
UV protection
response to ozone
response to lead ion
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
response to inactivity
hemoglobin metabolic process
negative regulation of NF-kappaB transcription factor activity
response to estradiol
response to insulin
response to vitamin A
response to vitamin E
response to L-ascorbic acid
response to drug
response to hydrogen peroxide
hydrogen peroxide catabolic process
negative regulation of apoptotic process
response to ethanol
response to cadmium ion
positive regulation of NF-kappaB transcription factor activity
protein tetramerization
protein homotetramerization
positive regulation of cell division
response to hyperoxia
response to fatty acid
cellular response to growth factor stimulus
response to phenylpropanoid
cellular oxidant detoxification
aminoacylase activity
catalase activity
receptor binding
antioxidant activity
oxidoreductase activity, acting on peroxide as acceptor
enzyme binding
heme binding
protein homodimerization activity
metal ion binding
NADP binding
response to reactive oxygen species
osteoblast differentiation
ureteric bud development
response to hypoxia
purine nucleotide catabolic process
triglyceride metabolic process
aging
cholesterol metabolic process
aerobic respiration
response to light intensity
UV protection
response to ozone
response to lead ion
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
response to inactivity
hemoglobin metabolic process
negative regulation of NF-kappaB transcription factor activity
response to estradiol
response to insulin
response to vitamin A
response to vitamin E
response to L-ascorbic acid
response to drug
response to hydrogen peroxide
hydrogen peroxide catabolic process
negative regulation of apoptotic process
response to ethanol
response to cadmium ion
positive regulation of NF-kappaB transcription factor activity
protein tetramerization
protein homotetramerization
positive regulation of cell division
response to hyperoxia
response to fatty acid
cellular response to growth factor stimulus
response to phenylpropanoid
cellular oxidant detoxification
aminoacylase activity
catalase activity
receptor binding
antioxidant activity
oxidoreductase activity, acting on peroxide as acceptor
enzyme binding
heme binding
protein homodimerization activity
metal ion binding
NADP binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11004185.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004185.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004185.hg.1 | TC11000327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004184.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004184.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004184.hg.1 | TC11000327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004183.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004183.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004183.hg.1 | TC11000327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000528104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000528104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000528104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151148.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151148.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151148.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151148.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151148.hg.1 | TC1100007257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100151150.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151150.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151150.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151150.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151150.hg.1 | TC1100007257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100151147.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151147.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151147.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151147.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151147.hg.1 | TC1100007257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100151149.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151149.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151149.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100151149.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100151149.hg.1 | TC1100007257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000241052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000525707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000528104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000528104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000528104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000530343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | ENST00000534710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007257.hg.1 | NM_001752.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 211100 | - | - |

CAT
Genomic Map