Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019786.hg.1 |
ENST00000308064.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019786.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019786.hg.1 |
TC11001589.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019787.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019787.hg.1 |
TC11001589.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019784.hg.1 |
ENST00000308064.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019784.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019784.hg.1 |
TC11001589.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019785.hg.1 |
ENST00000308064.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019785.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019785.hg.1 |
TC11001589.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180633.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180633.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180633.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180633.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180633.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180633.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180633.hg.1 |
TC1100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
ENST00000533673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010661.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180635.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180635.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180635.hg.1 |
TC1100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100180631.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180631.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180631.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180631.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180631.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180631.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180631.hg.1 |
TC1100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100180632.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180632.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180632.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180632.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180632.hg.1 |
ENST00000531322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180632.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180632.hg.1 |
TC1100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100180634.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180634.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180634.hg.1 |
ENST00000308064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100180634.hg.1 |
NM_003654.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100180634.hg.1 |
TC1100010661.hg.1 |
Clariom D TC |