Assay Details
| Gene Symbol: ZDHHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016353.4 | NP_057437.1 | 3 - 4 | 655 | 138 | |
| XM_011544548.2 | XP_011542850.1 | 2 - 3 | 121 | 138 | ||
| GenBank mRNA | AB023584.1 | - | 3 - 4 | 655 | 138 | |
| BC039253.1 | - | 3 - 4 | 460 | 5299722 | 138 | |
| BC050272.1 | - | 3 - 4 | 953 | 4398300 | 138 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000125.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011544548.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000517334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000517334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000517334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000523132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | ENST00000523132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001131.hg.1 | TC08000125.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001131.hg.1 | ENST00000262096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001131.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146415.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146415.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146415.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146415.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146415.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146415.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146415.hg.1 | TC0800006864.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000262096.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000517334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000517334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000517334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000522184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000523132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | ENST00000523132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006864.hg.1 | NM_016353.4 | Refseq |

ZDHHC2
Genomic Map