Assay Details
| Gene Symbol: ADGRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297704.1 | NP_001284633.1 | 9 - 10 | 1957 | 83 | |
| NM_001297705.1 | NP_001284634.1 | 8 - 9 | 1992 | 83 | ||
| NM_001297706.1 | NP_001284635.1 | 8 - 9 | 1992 | 83 | ||
| NM_012302.3 | NP_036434.1 | 8 - 9 | 1992 | 83 | ||
| XM_005270668.2 | XP_005270725.1 | 8 - 9 | 2009 | 83 | ||
| XM_017000788.1 | XP_016856277.1 | 10 - 11 | 1950 | 83 | ||
| XM_017000789.1 | XP_016856278.1 | 9 - 10 | 1934 | 83 | ||
| XM_017000792.1 | XP_016856281.1 | 9 - 10 | 1948 | 83 | ||
| XM_017000797.1 | XP_016856286.1 | 8 - 9 | 1918 | 83 | ||
| XM_017000798.1 | XP_016856287.1 | 8 - 9 | 1980 | 83 | ||
| GenBank mRNA | AF104266.1 | - | 8 - 9 | 1992 | 83 | |
| AF104938.1 | - | 8 - 9 | 1992 | 83 | ||
| AF104939.1 | - | 8 - 9 | 1992 | 83 | ||
| AJ131581.1 | - | 9 - 10 | 1956 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000800.hg.1
Set Membership: |
|

ADGRL2
Genomic Map