Assay Details
| Gene Symbol: MRPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145644.2 | NP_663619.1 | 4 - 5 | 572 | 65 | |
| GenBank mRNA | BC020651.1 | - | 4 - 5 | 548 | 4709909 | 65 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000522.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02008132.hg.1 | TC02000522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008132.hg.1 | ENST00000254644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008132.hg.1 | ENST00000337109.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008132.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008132.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008133.hg.1 | TC02000522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008133.hg.1 | ENST00000337109.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008133.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008133.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157572.hg.1 | TC0200008285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200157573.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157573.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157573.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157573.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157573.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157573.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157573.hg.1 | TC0200008285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000254644.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000337109.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000409180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | ENST00000605125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_016622.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_145644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008285.hg.1 | NM_145644.2 | Refseq |

MRPL35
Genomic Map