Assay Details
Gene Symbol: TEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003215.2 | NP_003206.2 | 10 - 11 | 959 | 137 | |
XM_011513737.2 | XP_011512039.1 | 10 - 11 | 940 | 137 | ||
XM_011513741.1 | XP_011512043.1 | 9 - 10 | 913 | 137 | ||
XM_017008569.1 | XP_016864058.1 | 9 - 10 | 2165 | 137 | ||
GenBank mRNA | BC101711.1 | - | 10 - 11 | 967 | 8069217 | 137 |
BC101713.1 | - | 10 - 11 | 967 | 8069219 | 137 | |
BC143487.1 | - | 10 - 11 | 967 | 9051995 | 137 | |
D29767.1 | - | 10 - 11 | 985 | 137 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001176.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
regulation of platelet activation
peptidyl-tyrosine phosphorylation
cell differentiation
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
Fc-epsilon receptor signaling pathway
regulation of cell proliferation
tissue regeneration
innate immune response
positive regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
phospholipid binding
metal ion binding
adaptive immune response
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
regulation of platelet activation
peptidyl-tyrosine phosphorylation
cell differentiation
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
Fc-epsilon receptor signaling pathway
regulation of cell proliferation
tissue regeneration
innate immune response
positive regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
phospholipid binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04016169.hg.1 | TC04001176.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016169.hg.1 | ENST00000381501.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016169.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016169.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172462.hg.1 | TC0400010618.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
