Assay Details
| Gene Symbol: VAMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185183.1 | NP_001172112.1 | 5 - 6 | 618 | 86 | |
| XM_011531188.1 | XP_011529490.1 | 5 - 6 | 597 | 86 | ||
| GenBank mRNA | AJ295938.1 | - | 5 - 6 | 530 | 86 | |
More Information
Set Membership: |
|

VAMP7
Genomic Map