Assay Details
| Gene Symbol: RPL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320138.1 | NP_001307067.1 | 1 - 2 | 274 | 116 | |
| GenBank mRNA | BP308263.1 | - | 1 - 2 | 240 | 116 | |
| CD688935.1 | - | 1 - 2 | 116 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320138.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
regulation of transcription, DNA-templated
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
cell-cell adhesion
DNA binding
RNA binding
structural constituent of ribosome
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
regulation of transcription, DNA-templated
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
cell-cell adhesion
DNA binding
RNA binding
structural constituent of ribosome
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000202773.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000424576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000546368.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000548343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000549847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000550566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000551291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000552455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | ENST00000553213.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_000970.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011971.hg.1 | NM_001024662.2 | Refseq |

RPL6
Genomic Map