Assay Details
Gene Symbol: BRD7P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002730.2 | 1 | 1320 | 94 | ||
GenBank mRNA | BC033937.1 | - | 1 | 74 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 85 like | CEP85L |
C6orf204, NY-BR-15, bA57K17.2 |
Anti-sense | Chr. 6 - 118460772 - 118710101 on NCBI Build GRCh38 | |
Downstream | phospholamban | PLN |
CMD1P, CMH18, PLB |
44.9kb | Sense | Chr. 6 - 118548263 - 118561715 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313660 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3847 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3846 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P42020 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 2138 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60391 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313660 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00317616 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3313660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217973 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217982 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217993 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15855516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217972 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05017105 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05017097 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05017095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217990 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15855516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05017097 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217990 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217972 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05017105 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05017095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217973 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217982 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25217993 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P42020 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P42020 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017083 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15855508 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017097 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017093 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15855514 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017081 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15855516 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017090 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017099 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15855501 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017095 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017088 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017086 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05017085 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15855503 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8121726 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8121728 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8121727 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8121725 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1569960_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 221093_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11753699_s_at | - | - |
Affymetrix Human Genome U219 Array | 11753699_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11763937_at | - | - |
Affymetrix Human Genome U219 Array | 11755454_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11763937_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11755454_s_at | - | - |
Affymetrix Human Clariom D Assay | PSR0600163657.hg.1 | NR_002730.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600163657.hg.1 | NR_002730.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600163657.hg.1 | TC0600009268.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1569960_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1569960_at | - | - |
Human Genome U133 Plus 2.0 Array | 221093_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 221093_at | - | - |
Human Genome U133A 2.0 Array | 221093_at | - | - |
Human Genome U133A 2.0 Array | 221093_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002730_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw516181 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569960_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221093_at11 | - | - |
Human Genome U133 Set | 221093_PM_at | - | - |
Human Genome U133 Set | 221093_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032007 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032006 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032006 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032007 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 202289 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2147920 | NR_002730.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2147920 | AF167351.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3245301 | NR_002730.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3245301 | BC033937.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2147920 | NR_002730.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2147920 | AF167351.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3245301 | NR_002730.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3245301 | BC033937.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2147920 | NR_002730.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2147920 | AF167351.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245301 | NR_002730.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245301 | BC033937.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2147920 | NR_002730.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2147920 | AF167351.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245301 | NR_002730.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245301 | BC033937.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2147920 | NR_002730.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2147920 | AF167351.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2147920 | NR_002730.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2147920 | AF167351.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2147920 | NR_002730.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2147920 | AF167351.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2147920 | NR_002730.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2147920 | AF167351.1 | human_transcriptome |
