Assay Details
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256409.1 | NP_001243338.1 | 6 - 7 | 1084 | 124 | |
NM_052940.4 | NP_443172.1 | 5 - 6 | 1050 | 124 | ||
XM_006710327.3 | XP_006710390.1 | 6 - 7 | 906 | 124 | ||
XM_006710328.3 | XP_006710391.1 | 6 - 7 | 1419 | 124 | ||
GenBank mRNA | AK074521.1 | - | 5 - 6 | 1023 | 124 | |
AK075201.1 | - | 5 - 6 | 1012 | 124 | ||
AK090881.1 | - | 6 - 7 | 1333 | 124 | ||
AK095767.1 | - | 6 - 7 | 1003 | 124 | ||
BC013101.1 | - | 5 - 6 | 889 | 3918785 | 124 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000659.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01010715.hg.1 | TC01000659.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01010715.hg.1 | ENST00000319223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010715.hg.1 | ENST00000371370.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010715.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01010715.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01010715.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159614.hg.1 | TC0100008369.hg.1 | Clariom D TC |
