Assay Details
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006710328.3 | XP_006710391.1 | 1 - 2 | 466 | 156 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006710328.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
