Assay Details
| Gene Symbol: BET1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016526.4 | NP_057610.2 | 3 | 390 | 74 | |
| GenBank mRNA | BC032779.2 | - | 3 | 350 | 4938985 | 74 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BET1L
Genomic Map