Assay Details
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016231.4 | NP_057315.3 | 2 - 3 | 807 | 79 | |
XM_005257988.2 | XP_005258045.1 | 2 - 3 | 822 | 79 | ||
GenBank mRNA | AF180819.1 | - | 2 - 3 | 805 | 79 | |
AF197898.1 | - | 2 - 3 | 807 | 79 | ||
AK024686.1 | - | 2 - 3 | 439 | 79 | ||
AK315315.1 | - | 2 - 3 | 822 | 79 | ||
BC064663.1 | - | 2 - 3 | 1319 | 6527673 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000315.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
transforming growth factor beta receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
peptidyl-threonine phosphorylation
negative regulation of Wnt signaling pathway
serine phosphorylation of STAT3 protein
intracellular signal transduction
protein autophosphorylation
protein stabilization
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
ubiquitin protein ligase binding
SH2 domain binding
MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
transforming growth factor beta receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
peptidyl-threonine phosphorylation
negative regulation of Wnt signaling pathway
serine phosphorylation of STAT3 protein
intracellular signal transduction
protein autophosphorylation
protein stabilization
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
ubiquitin protein ligase binding
SH2 domain binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17003806.hg.1 | ENST00000407008.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17003806.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003806.hg.1 | TC17000315.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000583517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000583517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000583517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152209.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152209.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152209.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152209.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152209.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152209.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152209.hg.1 | TC1700007344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000407008.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000496808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000582037.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000583517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000583517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000583517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | ENST00000584878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007344.hg.1 | NM_016231.4 | Refseq |
