Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003043.hg.1 |
TC08000209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003043.hg.1 |
ENST00000337221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003043.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003043.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003045.hg.1 |
TC08000209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003045.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003045.hg.1 |
ENST00000337221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003045.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003045.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003046.hg.1 |
TC08000209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003046.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003046.hg.1 |
ENST00000337221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003046.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003046.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003044.hg.1 |
TC08000209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003044.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003044.hg.1 |
ENST00000337221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003044.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003044.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148846.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148846.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148846.hg.1 |
TC0800007114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148850.hg.1 |
TC0800007114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800148847.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148847.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148847.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148847.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148847.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148847.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148847.hg.1 |
TC0800007114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148851.hg.1 |
TC0800007114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000301904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
ENST00000337221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_016240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007114.hg.1 |
NM_182826.1 |
Refseq |