Assay Details
| Gene Symbol: TOMM40L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286373.1 | NP_001273302.1 | 1 - 2 | 199 | 106 | |
| NM_001286374.1 | NP_001273303.1 | 1 - 2 | 199 | 106 | ||
| NM_032174.5 | NP_115550.2 | 1 - 2 | 199 | 106 | ||
| GenBank mRNA | AK022832.1 | - | 1 - 2 | 78 | 106 | |
| AK027704.1 | - | 1 - 2 | 78 | 106 | ||
| AK297848.1 | - | 1 - 2 | 78 | 106 | ||
| BX537951.1 | - | 1 - 2 | 76 | 106 | ||
| DA160426.1 | - | 1 - 2 | 95 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK022832.1, AK027704.1, AK297848.1, BX537951.1, NM_001286373.1, NM_001286374.1, NM_032174.5 transcripts.
Set Membership: |
|

TOMM40L
Genomic Map