Assay Details
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006135.2 | NP_006126.1 | 6 - 7 | 901 | 67 | |
XM_011542225.2 | XP_011540527.1 | 6 - 7 | 645 | 67 | ||
XM_017002424.1 | XP_016857913.1 | 6 - 7 | 645 | 67 | ||
GenBank mRNA | AK223226.1 | - | 6 - 7 | 522 | 67 | |
AK289654.1 | - | 6 - 7 | 544 | 67 | ||
BC000144.1 | - | 6 - 7 | 526 | 3047937 | 67 | |
BT019364.1 | - | 6 - 7 | 509 | 67 | ||
BX648738.1 | - | 6 - 7 | 1192 | 67 | ||
CR407657.1 | - | 6 - 7 | 509 | 67 | ||
CR541819.1 | - | 6 - 7 | 509 | 67 | ||
U56637.1 | - | 6 - 7 | 509 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000990.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01015747.hg.1 | TC01000990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015747.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015747.hg.1 | ENST00000476936.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015747.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01015753.hg.1 | TC01000990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015753.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015753.hg.1 | ENST00000476936.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015753.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01015755.hg.1 | TC01000990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015755.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015755.hg.1 | ENST00000476936.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015755.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01015752.hg.1 | TC01000990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015752.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015752.hg.1 | ENST00000476936.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015752.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012645.hg.1 | ENST00000418056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012645.hg.1 | ENST00000418056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015749.hg.1 | TC01000990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015749.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015749.hg.1 | ENST00000476936.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015749.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167981.hg.1 | TC0100009444.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167983.hg.1 | TC0100009444.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000485542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | ENST00000498626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009444.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167987.hg.1 | TC0100009444.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012645.hg.1 | ENST00000418056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012645.hg.1 | ENST00000418056.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000476820.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167986.hg.1 | TC0100009444.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | ENST00000263168.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | ENST00000466066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | ENST00000476936.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | NM_006135.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167989.hg.1 | TC0100009444.hg.1 | Clariom D TC |
