Assay Details
| Gene Symbol: POLDIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015584.4 | NP_056399.1 | 2 - 3 | 380 | 76 | |
| GenBank mRNA | AF077203.1 | - | 2 - 3 | 288 | 76 | |
| AF179891.1 | - | 2 - 3 | 239 | 76 | ||
| AK313232.1 | - | 2 - 3 | 272 | 76 | ||
| BC000655.2 | - | 2 - 3 | 272 | 3349399 | 76 | |
| BC009265.2 | - | 2 - 3 | 266 | 3943941 | 76 | |
| BC018864.2 | - | 2 - 3 | 305 | 76 | ||
| BU932070.1 | - | 2 - 3 | 334 | 76 | ||
| DB455962.1 | - | 2 - 3 | 371 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001295.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17017096.hg.1 | ENST00000003607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017096.hg.1 | ENST00000540200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017096.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017096.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017097.hg.1 | ENST00000003607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017097.hg.1 | ENST00000540200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017097.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017097.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179679.hg.1 | TC1700010191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179680.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179680.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179680.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179680.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179680.hg.1 | TC1700010191.hg.1 | Clariom D TC |

POLDIP2
Genomic Map