Assay Details
| Gene Symbol: BAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001188.3 | NP_001179.1 | 1 - 2 | 271 | 71 | |
| XM_011514780.1 | XP_011513082.1 | 1 - 2 | 254 | 71 | ||
| GenBank mRNA | AK091807.1 | - | 1 - 2 | 198 | 71 | |
| AK303422.1 | - | 1 - 2 | 250 | 71 | ||
| BC004431.1 | - | 1 | 182 | 2819507 | 71 | |
| BC110337.1 | - | 1 - 2 | 182 | 5755200 | 70 | |
| CB142221.1 | - | 1 - 2 | 271 | 71 | ||
| U16811.1 | - | 1 - 2 | 171 | 71 | ||
| U23765.1 | - | 1 - 2 | 35 | 71 | ||
| X84213.1 | - | 1 - 2 | 163 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK091807.1, AK303422.1, BC004431.1, BC110337.1, NM_001188.3, U16811.1, U23765.1, X84213.1, XM_011514780.1 transcripts.
Set Membership: |
|

BAK1
Genomic Map