Assay Details
Gene Symbol: MAP3K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005921.1 | NP_005912.1 | 16 - 17 | 4484 | 131 | |
XM_017009484.1 | XP_016864973.1 | 16 - 17 | 7195 | 131 | ||
XM_017009485.1 | XP_016864974.1 | 16 - 17 | 3923 | 131 | ||
GenBank mRNA | AF042838.1 | - | 16 - 17 | 3932 | 131 | |
BU194120.1 | - | 3 - 4 | 443 | 131 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000243.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
MyD88-dependent toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
Fc-epsilon receptor signaling pathway
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
zinc ion binding
protein kinase binding
MAPK cascade
activation of MAPKK activity
MyD88-dependent toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
Fc-epsilon receptor signaling pathway
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
zinc ion binding
protein kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05003004.hg.1 | TC05000243.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05003004.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05003004.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05003004.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150357.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150357.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150357.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150357.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150357.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150357.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150357.hg.1 | TC0500007452.hg.1 | Clariom D TC |
