Assay Details
Gene Symbol: MAP3K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005921.1 | NP_005912.1 | 17 - 18 | 4620 | 116 | |
XM_017009484.1 | XP_016864973.1 | 17 - 18 | 7331 | 116 | ||
XM_017009485.1 | XP_016864974.1 | 17 - 18 | 4059 | 116 | ||
GenBank mRNA | BU194120.1 | - | 4 - 5 | 579 | 116 | |
BX537527.1 | - | 1 - 2 | 116 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000243.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
MyD88-dependent toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
Fc-epsilon receptor signaling pathway
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
zinc ion binding
protein kinase binding
MAPK cascade
activation of MAPKK activity
MyD88-dependent toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
Fc-epsilon receptor signaling pathway
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
zinc ion binding
protein kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05003005.hg.1 | TC05000243.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05003005.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05003005.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05003005.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150358.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150358.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150358.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150358.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150358.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150358.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150358.hg.1 | TC0500007452.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
