Assay Details
Gene Symbol: MAP3K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005921.1 | NP_005912.1 | 6 - 7 | 1804 | 120 | |
XM_017009484.1 | XP_016864973.1 | 6 - 7 | 4515 | 120 | ||
XM_017009485.1 | XP_016864974.1 | 6 - 7 | 1243 | 120 | ||
GenBank mRNA | AF042838.1 | - | 6 - 7 | 1246 | 120 | |
AK307828.1 | - | 7 - 8 | 1024 | 120 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000243.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
MyD88-dependent toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
Fc-epsilon receptor signaling pathway
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
zinc ion binding
protein kinase binding
MAPK cascade
activation of MAPKK activity
MyD88-dependent toll-like receptor signaling pathway
protein phosphorylation
apoptotic process
Fc-epsilon receptor signaling pathway
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
zinc ion binding
protein kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05002994.hg.1 | TC05000243.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05002994.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05002994.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05002993.hg.1 | TC05000243.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05002993.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05002993.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | ENST00000469188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007452.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150347.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150347.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150347.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150347.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150347.hg.1 | TC0500007452.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500150346.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150346.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150346.hg.1 | ENST00000399503.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500150346.hg.1 | NM_005921.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500150346.hg.1 | TC0500007452.hg.1 | Clariom D TC |
