Assay Details
| Gene Symbol: SLC37A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320537.1 | NP_001307466.1 | 17 - 18 | 2473 | 121 | |
| NM_018964.3 | NP_061837.3 | 18 - 19 | 1842 | 121 | ||
| XM_011529614.2 | XP_011527916.1 | 16 - 17 | 2614 | 121 | ||
| XM_011529615.1 | XP_011527917.1 | 16 - 17 | 2614 | 121 | ||
| XM_017028376.1 | XP_016883865.1 | 16 - 17 | 2614 | 121 | ||
| XM_017028377.1 | XP_016883866.1 | 18 - 19 | 1767 | 121 | ||
| XM_017028378.1 | XP_016883867.1 | 18 - 19 | 2226 | 121 | ||
| XM_017028379.1 | XP_016883868.1 | 19 - 20 | 1873 | 121 | ||
| XM_017028380.1 | XP_016883869.1 | 17 - 18 | 1673 | 121 | ||
| XM_017028381.1 | XP_016883870.1 | 18 - 19 | 1767 | 121 | ||
| XM_017028382.1 | XP_016883871.1 | 16 - 17 | 2955 | 121 | ||
| GenBank mRNA | AF311320.1 | - | 17 - 18 | 2412 | 121 | |
| AJ269529.2 | - | 18 - 19 | 1842 | 121 | ||
| AK025993.1 | - | 4 - 5 | 305 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000202.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002114.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002114.hg.1 | ENST00000398341.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002114.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002114.hg.1 | TC21000202.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | TC2100008527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158353.hg.1 | NM_018964.3 | Refseq |

SLC37A1
Genomic Map