Assay Details
Gene Symbol: SLC37A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320537.1 | NP_001307466.1 | 4 - 5 | 1306 | 64 | |
NM_018964.3 | NP_061837.3 | 5 - 6 | 675 | 64 | ||
XM_011529614.2 | XP_011527916.1 | 3 - 4 | 1447 | 64 | ||
XM_011529615.1 | XP_011527917.1 | 3 - 4 | 1447 | 64 | ||
XM_017028376.1 | XP_016883865.1 | 3 - 4 | 1447 | 64 | ||
XM_017028377.1 | XP_016883866.1 | 5 - 6 | 600 | 64 | ||
XM_017028378.1 | XP_016883867.1 | 5 - 6 | 1059 | 64 | ||
XM_017028379.1 | XP_016883868.1 | 6 - 7 | 706 | 64 | ||
XM_017028380.1 | XP_016883869.1 | 4 - 5 | 506 | 64 | ||
XM_017028381.1 | XP_016883870.1 | 5 - 6 | 600 | 64 | ||
XM_017028382.1 | XP_016883871.1 | 3 - 4 | 1788 | 64 | ||
XM_017028383.1 | XP_016883872.1 | 5 - 6 | 531 | 64 | ||
GenBank mRNA | AF311320.1 | - | 4 - 5 | 1245 | 64 | |
AJ269529.2 | - | 5 - 6 | 675 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000202.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR21002100.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002100.hg.1 | ENST00000398341.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002100.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002100.hg.1 | TC21000202.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002099.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002099.hg.1 | ENST00000398341.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002099.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002099.hg.1 | TC21000202.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2100158338.hg.1 | TC2100008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100158338.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158338.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158338.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158338.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158338.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158338.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158339.hg.1 | TC2100008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100158339.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158339.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158339.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158339.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158339.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158339.hg.1 | NM_018964.3 | Refseq |
