Assay Details
| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013437.4 | NP_038465.1 | 2 - 3 | 260 | 83 | |
| GenBank mRNA | AF166350.1 | - | 2 - 3 | 175 | 83 | |
| AK289752.1 | - | 2 - 3 | 164 | 83 | ||
| AL698796.1 | - | 2 - 3 | 228 | 83 | ||
| BC032109.2 | - | 2 - 3 | 136 | 4842797 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001511.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08019873.hg.1 | TC08001511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019873.hg.1 | ENST00000276654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019873.hg.1 | ENST00000424843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019873.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019873.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177707.hg.1 | TC0800011395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |

LRP12
Genomic Map