Assay Details
Gene Symbol: BFSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161705.1 | NP_001155177.1 | 5 - 6 | 628 | 147 | |
NM_001195.4 | NP_001186.1 | 5 - 6 | 772 | 147 | ||
NM_001278606.1 | NP_001265535.1 | 6 - 7 | 763 | 147 | ||
NM_001278607.1 | NP_001265536.1 | 5 - 6 | 624 | 147 | ||
NM_001278608.1 | NP_001265537.1 | 7 - 8 | 749 | 147 | ||
GenBank mRNA | AF039655.1 | - | 5 - 6 | 732 | 147 | |
AK304673.1 | - | 7 - 8 | 749 | 147 | ||
AK316247.1 | - | 6 - 7 | 641 | 147 | ||
BC041483.1 | - | 5 - 6 | 628 | 5406467 | 147 | |
BQ230198.1 | - | 5 - 6 | 467 | 147 | ||
Y16717.2 | - | 5 - 6 | 732 | 147 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000655.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20009175.hg.1 | ENST00000377868.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20009175.hg.1 | ENST00000377873.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20009175.hg.1 | ENST00000536626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20009175.hg.1 | ENST00000544874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20009175.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20009175.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20009175.hg.1 | TC20000655.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | TC2000008493.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000159602.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
