Assay Details
| Gene Symbol: TK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172645.1 | NP_001166116.1 | 1 - 2 | 475 | 124 | |
| NM_004614.4 | NP_004605.4 | 1 - 2 | 475 | 124 | ||
| GenBank mRNA | AF521891.1 | - | 1 - 2 | 475 | 124 | |
| AK294627.1 | - | 1 - 2 | 124 | |||
| AK303590.1 | - | 1 - 2 | 253 | 124 | ||
| Y10498.3 | - | 1 - 2 | 240 | 124 | ||
More Information
Set Membership: |
|

TK2
Genomic Map