Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024208.hg.1 |
TC06001964.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024208.hg.1 |
ENST00000369239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024208.hg.1 |
ENST00000438806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024208.hg.1 |
NM_015491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024208.hg.1 |
NM_032870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000369239.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000438806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000460600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000476159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000478777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
NM_015491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
NM_032870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000369239.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000438806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000460600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000476159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
ENST00000478777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
NM_015491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
NM_032870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187186.hg.1 |
TC0600012655.hg.1 |
Clariom D TC |