| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012016.hg.1 |
ENST00000375735.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012016.hg.1 |
ENST00000398093.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012016.hg.1 |
ENST00000540621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012016.hg.1 |
NM_001080463.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012016.hg.1 |
NM_001377.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012016.hg.1 |
TC11000963.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000375735.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000398093.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000528670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000533197.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
NM_001080463.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
NM_001377.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000375735.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000398093.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000528670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
ENST00000533197.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
NM_001080463.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
NM_001377.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100165940.hg.1 |
TC1100008927.hg.1 |
Clariom D TC |