Assay Details
Gene Symbol: SYNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033071.3 | NP_149062.1 | 127 - 128 | 23526 | 57 | |
NM_182961.3 | NP_892006.3 | 128 - 129 | 23904 | 57 | ||
XM_006715407.1 | XP_006715470.1 | 128 - 129 | 23817 | 57 | ||
XM_006715408.2 | XP_006715471.1 | 128 - 129 | 23809 | 57 | ||
XM_006715409.1 | XP_006715472.1 | 127 - 128 | 23796 | 57 | ||
XM_006715410.2 | XP_006715473.1 | 128 - 129 | 23821 | 57 | ||
XM_006715411.1 | XP_006715474.1 | 127 - 128 | 23766 | 57 | ||
XM_006715412.2 | XP_006715475.1 | 128 - 129 | 23806 | 57 | ||
XM_006715413.2 | XP_006715476.1 | 128 - 129 | 23821 | 57 | ||
XM_006715414.1 | XP_006715477.1 | 126 - 127 | 23745 | 57 | ||
XM_006715415.2 | XP_006715478.1 | 128 - 129 | 23821 | 57 | ||
XM_006715416.2 | XP_006715479.1 | 128 - 129 | 23806 | 57 | ||
XM_006715417.2 | XP_006715480.1 | 127 - 128 | 23680 | 57 | ||
XM_006715420.2 | XP_006715483.1 | 127 - 128 | 23668 | 57 | ||
XM_006715421.2 | XP_006715484.1 | 127 - 128 | 23665 | 57 | ||
XM_006715422.1 | XP_006715485.1 | 127 - 128 | 23658 | 57 | ||
XM_006715423.2 | XP_006715486.1 | 128 - 129 | 23821 | 57 | ||
XM_006715424.2 | XP_006715487.1 | 128 - 129 | 23821 | 57 | ||
XM_006715425.2 | XP_006715488.1 | 128 - 129 | 23821 | 57 | ||
XM_011535641.2 | XP_011533943.1 | 128 - 129 | 23818 | 57 | ||
XM_011535642.2 | XP_011533944.1 | 128 - 129 | 23806 | 57 | ||
XM_011535643.1 | XP_011533945.1 | 127 - 128 | 23652 | 57 | ||
XM_011535644.1 | XP_011533946.1 | 114 - 115 | 21868 | 57 | ||
XM_011535645.2 | XP_011533947.1 | 110 - 111 | 21261 | 57 | ||
XM_017010608.1 | XP_016866097.1 | 129 - 130 | 23936 | 57 | ||
XM_017010609.1 | XP_016866098.1 | 128 - 129 | 23743 | 57 | ||
XM_017010610.1 | XP_016866099.1 | 127 - 128 | 23722 | 57 | ||
XM_017010611.1 | XP_016866100.1 | 127 - 128 | 23525 | 57 | ||
XM_017010612.1 | XP_016866101.1 | 126 - 127 | 23449 | 57 | ||
XM_017010613.1 | XP_016866102.1 | 128 - 129 | 23818 | 57 | ||
XM_017010614.1 | XP_016866103.1 | 127 - 128 | 23665 | 57 | ||
XM_017010615.1 | XP_016866104.1 | 127 - 128 | 23665 | 57 | ||
XM_017010616.1 | XP_016866105.1 | 128 - 129 | 23821 | 57 | ||
XM_017010617.1 | XP_016866106.1 | 128 - 129 | 23818 | 57 | ||
XM_017010618.1 | XP_016866107.1 | 128 - 129 | 23806 | 57 | ||
XM_017010619.1 | XP_016866108.1 | 114 - 115 | 21900 | 57 | ||
GenBank mRNA | AB018339.2 | - | 16 - 17 | 2541 | 57 | |
AF444779.1 | - | 3 - 4 | 414 | 57 | ||
AF495910.1 | - | 128 - 129 | 23904 | 57 | ||
AF535142.1 | - | 127 - 128 | 23526 | 57 | ||
AK122691.1 | - | 8 - 9 | 1184 | 57 | ||
AK127076.1 | - | 12 - 13 | 1881 | 57 | ||
AK316101.1 | - | 1 - 2 | 121 | 57 | ||
AY061755.1 | - | 43 - 44 | 6994 | 57 | ||
AY184206.1 | - | 41 - 42 | 6889 | 57 | ||
BC090927.1 | - | 1 - 2 | 166 | 57 | ||
BX537837.1 | - | 10 - 11 | 1482 | 57 | ||
BX647837.1 | - | 13 - 14 | 2076 | 57 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002238.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK316101.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
