Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
TC06002238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000265368.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000341594.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000347037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000354674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000356820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000367247.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000367255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000423061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000448038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
ENST00000539504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
NM_033071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027921.hg.1 |
NM_182961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000341594.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000347037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000354674.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367257.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000409694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000423061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000460912.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000478916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000536990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000539504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
NM_033071.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
NM_182961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000341594.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000347037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000354674.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367256.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000367257.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000409694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000423061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000460912.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000478916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000536990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
ENST00000539504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
NM_033071.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
NM_182961.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193766.hg.1 |
TC0600013588.hg.1 |
Clariom D TC |