Assay Details
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018066.3 | NP_060536.3 | 3 - 4 | 1010 | 135 | |
GenBank mRNA | AK001211.1 | - | 3 - 4 | 929 | 135 | |
AK022535.1 | - | 3 - 4 | 915 | 135 | ||
AY358864.1 | - | 3 - 4 | 919 | 135 | ||
AY364261.1 | - | 3 - 4 | 728 | 135 | ||
BC007815.2 | - | 3 - 4 | 852 | 4299794 | 135 | |
BC008634.1 | - | 3 - 4 | 876 | 3463813 | 135 | |
BC020272.1 | - | 3 - 4 | 895 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002395.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01036964.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036964.hg.1 | ENST00000374131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036964.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036964.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036964.hg.1 | ENST00000477418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036964.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036966.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036966.hg.1 | ENST00000374131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036966.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036966.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036966.hg.1 | ENST00000477418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036966.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199583.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199585.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199584.hg.1 | TC0100013408.hg.1 | Clariom D TC |
