Assay Details
Gene Symbol: TOMM40L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286373.1 | NP_001273302.1 | 4 - 5 | 613 | 79 | |
XM_017002482.1 | XP_016857971.1 | 4 - 5 | 481 | 79 | ||
GenBank mRNA | AK297848.1 | - | 4 - 5 | 492 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
