Assay Details
Gene Symbol: RAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013435.2 | NP_038463.2 | 1 - 2 | 480 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000545.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
pattern specification process
visual perception
hypothalamus development
camera-type eye development
positive regulation of transcription from RNA polymerase II promoter
limb development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription from RNA polymerase II promoter
pattern specification process
visual perception
hypothalamus development
camera-type eye development
positive regulation of transcription from RNA polymerase II promoter
limb development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18005661.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005661.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005661.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005661.hg.1 | TC18000545.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18005660.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005660.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005660.hg.1 | TC18000545.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000555288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000555288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000591550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | ENST00000591550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005662.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005662.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005662.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005662.hg.1 | TC18000545.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800160717.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160717.hg.1 | ENST00000591550.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160717.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160717.hg.1 | TC1800008821.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800160717.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160717.hg.1 | ENST00000591550.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160717.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000555288.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000555288.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000591550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | ENST00000591550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008821.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160724.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160724.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160724.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160724.hg.1 | TC1800008821.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800160724.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160724.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160724.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160720.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160720.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160720.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160720.hg.1 | TC1800008821.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800160720.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160720.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160720.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | ENST00000555288.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | NM_013435.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | TC1800008821.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | ENST00000256852.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | ENST00000334889.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | ENST00000555288.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160721.hg.1 | NM_013435.2 | Refseq |
