Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023644.hg.1 |
TC06001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023644.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023644.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186211.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186211.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186211.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186211.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186211.hg.1 |
TC0600012514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000257787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
ENST00000420494.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012514.hg.1 |
NM_018064.3 |
Refseq |