Assay Details
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017946.3 | NP_060416.1 | 2 - 3 | 637 | 104 | |
GenBank mRNA | AK000738.1 | - | 2 - 3 | 489 | 104 | |
AY358643.1 | - | 2 - 3 | 517 | 104 | ||
BC005206.1 | - | 2 - 3 | 520 | 4042173 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001245.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07019395.hg.1 | TC07001245.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07019395.hg.1 | ENST00000222803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07019395.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | ENST00000479939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010621.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | ENST00000222803.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | ENST00000412494.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | ENST00000419018.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | NM_017946.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | NR_046478.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | NR_046479.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700175501.hg.1 | TC0700010621.hg.1 | Clariom D TC |
