Assay Details
Gene Symbol: SLC37A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018964.3 | NP_061837.3 | 1 - 2 | 106 | 77 | |
GenBank mRNA | AJ269529.2 | - | 1 - 2 | 106 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000202.hg
Additional Information:
This assay is in the 5'UTR region of the AJ269529.2,NM_018964.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002084.hg.1 | ENST00000398341.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002084.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002084.hg.1 | TC21000202.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21002085.hg.1 | ENST00000398341.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002085.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002085.hg.1 | TC21000202.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21002083.hg.1 | ENST00000398341.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21002083.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21002083.hg.1 | TC21000202.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2100158324.hg.1 | TC2100008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100158324.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158324.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158324.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158324.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158324.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158324.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158323.hg.1 | TC2100008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100158323.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158323.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158323.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158323.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158325.hg.1 | TC2100008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100158325.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158325.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158325.hg.1 | NM_018964.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158325.hg.1 | ENST00000398341.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158325.hg.1 | ENST00000484887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158325.hg.1 | NM_018964.3 | Refseq |
