Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
TC0X000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
ENST00000349874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
ENST00000359763.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
ENST00000361511.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
ENST00000543630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
ENST00000544635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
NM_006240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
NM_152224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001211.hg.1 |
NM_152226.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000349874.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000361511.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000379962.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000496075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
NM_006240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
NM_152224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
NM_152226.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000349874.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000361511.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000379962.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
ENST00000496075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
NM_006240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
NM_152224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
NM_152226.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146107.hg.1 |
TC0X00006725.hg.1 |
Clariom D TC |