Assay Details
| Gene Symbol: SGTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003021.3 | NP_003012.1 | 1 - 2 | 124 | 137 | |
| GenBank mRNA | BG423904.1 | - | 125 | 138 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001036.hg
Additional Information:
This assay is in the 5'UTR region of the NM_003021.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19014131.hg.1 | ENST00000221566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014131.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014131.hg.1 | TC19001036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014135.hg.1 | ENST00000221566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014135.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014135.hg.1 | TC19001036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000586711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014134.hg.1 | ENST00000221566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014134.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014134.hg.1 | TC19001036.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | TC1900009254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173746.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900173755.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173755.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900173755.hg.1 | TC1900009254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900173755.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900173755.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000221566.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000586711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000587866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000589251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | ENST00000591984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009254.hg.1 | NM_003021.3 | Refseq |

SGTA
Genomic Map