Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013375.hg.1 |
ENST00000378076.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013375.hg.1 |
ENST00000538044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013375.hg.1 |
NM_003638.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013375.hg.1 |
TC10001064.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000167569.hg.1 |
ENST00000378076.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167569.hg.1 |
NM_001291494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167569.hg.1 |
NM_003638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167569.hg.1 |
ENST00000378076.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167569.hg.1 |
NM_001291494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167569.hg.1 |
NM_003638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167569.hg.1 |
TC1000009893.hg.1 |
Clariom D TC |