Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013167.hg.1 |
TC01000827.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013167.hg.1 |
ENST00000356813.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013167.hg.1 |
ENST00000370548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013167.hg.1 |
ENST00000370550.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013167.hg.1 |
ENST00000370551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013167.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013167.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238260.hg.1 |
TC0100018242.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370550.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000370551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
ENST00000591456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_001134492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018242.hg.1 |
NM_012262.3 |
Refseq |