| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013711.hg.1 |
ENST00000447495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012875.hg.1 |
ENST00000433562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012875.hg.1 |
ENST00000433562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007757.hg.1 |
ENST00000414685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000517630.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000518360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520595.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000520809.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000521628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523111.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
ENST00000523432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007554.hg.1 |
NM_003350.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007757.hg.1 |
ENST00000414685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012875.hg.1 |
ENST00000433562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012875.hg.1 |
ENST00000433562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013711.hg.1 |
ENST00000447495.1 |
Ensembl |