Assay Details
| Gene Symbol: SLC13A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318192.1 | NP_001305121.1 | 12 - 13 | 2030 | 128 | |
| NM_012450.3 | NP_036582.2 | 12 - 13 | 2027 | 128 | ||
| XM_011516024.2 | XP_011514326.1 | 12 - 13 | 1810 | 128 | ||
| XM_017011962.1 | XP_016867451.1 | 10 - 11 | 1398 | 128 | ||
| GenBank mRNA | AB208809.1 | - | 9 - 10 | 6305 | 128 | |
| AF169301.1 | - | 12 - 13 | 2000 | 128 | ||
| AK313072.1 | - | 12 - 13 | 1620 | 128 | ||
| BC030689.2 | - | 12 - 13 | 2009 | 4821193 | 128 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001888.hg.1
Set Membership: |
|

SLC13A4
Genomic Map