Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037303.hg.1 |
TC03002109.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037303.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037303.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013522.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200886.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200886.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200886.hg.1 |
ENST00000354905.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200886.hg.1 |
NM_207316.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200886.hg.1 |
TC0300013522.hg.1 |
Clariom D TC |