Assay Details
| Gene Symbol: PRDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005809.5 | NP_005800.3 | 4 - 5 | 558 | 66 | |
| GenBank mRNA | AK293957.1 | - | 4 - 5 | 478 | 66 | |
| BC000452.2 | - | 4 - 5 | 437 | 2821457 | 66 | |
| BC003022.1 | - | 4 - 5 | 437 | 2821457 | 66 | |
| BC039428.1 | - | 4 - 5 | 497 | 5303023 | 66 | |
| CR450356.1 | - | 3 - 4 | 378 | 66 | ||
| CR541789.1 | - | 3 - 4 | 378 | 66 | ||
| EU668325.1 | - | 4 - 5 | 437 | 66 | ||
| L19185.1 | - | 4 - 5 | 502 | 66 | ||
| X82321.1 | - | 2 - 3 | 277 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001210.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
response to oxidative stress
removal of superoxide radicals
cellular response to oxidative stress
hydrogen peroxide catabolic process
regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
cell redox homeostasis
oxidation-reduction process
thioredoxin peroxidase activity
antioxidant activity
response to reactive oxygen species
response to oxidative stress
removal of superoxide radicals
cellular response to oxidative stress
hydrogen peroxide catabolic process
regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
cell redox homeostasis
oxidation-reduction process
thioredoxin peroxidase activity
antioxidant activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017005.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017005.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017005.hg.1 | TC19001210.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017001.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017001.hg.1 | ENST00000334482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017001.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017001.hg.1 | TC19001210.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007270.hg.1 | ENST00000451920.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007270.hg.1 | ENST00000451920.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007270.hg.1 | ENST00000451920.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017007.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017007.hg.1 | ENST00000334482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017007.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017007.hg.1 | TC19001210.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017004.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017004.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017004.hg.1 | TC19001210.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | TC1900009752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179638.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | TC1900009752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179641.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | TC1900009752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179639.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | TC1900009752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179635.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | TC1900009752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900179642.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007270.hg.1 | ENST00000451920.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007270.hg.1 | ENST00000451920.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007270.hg.1 | ENST00000451920.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000301522.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000334482.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000466174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000477555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000478908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | ENST00000498785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009752.hg.1 | NM_005809.5 | Refseq |

PRDX2
Genomic Map